1-Pyrrolidino-1-cyclohexene, 95%
1-Pyrrolidino-1-cyclohexene, 95%
1-Pyrrolidino-1-cyclohexene, 95%
Thermo Scientific Chemicals

1-Pyrrolidino-1-cyclohexene, 95%

CAS: 1125-99-1 | C10H17N | 151.25 g/mol
Quantity:
25 g
100 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 334831000
Price (JPY)
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS1125-99-1
IUPAC Name1-(cyclohex-1-en-1-yl)pyrrolidine
Molecular FormulaC10H17N
InChI KeyKTZNVZJECQAMBV-UHFFFAOYSA-N
SMILESC1CCN(C1)C1=CCCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.5200 to 1.5240 (20°C, 589 nm)
Infrared spectrumConforms
GC>=94 %
AppearanceMay darken on storage
Appearance (Color)Clear colorless to yellow-brown
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only