1-Step™ NBT/BCIP Substrate Solution
Thermo Scientific™

1-Step™ NBT/BCIP Substrate Solution

Thermo Scientific Pierce NBT and BCIP Substrates are powders and ready-made solutions of nitro-blue tetrazolium and 5-bromo-4-chloro-3'-indolyphosphate for chromogenic blot자세히 알아보기
Have Questions?
카탈로그 번호수량
34042250 mL
카탈로그 번호 34042
제품 가격(KRW)
261,000
온라인 행사
Ends: 31-Dec-2026
289,000
할인액 28,000 (10%)
Each
수량:
250 mL
대량 주문 또는 맞춤형 요청
제품 가격(KRW)
261,000
온라인 행사
Ends: 31-Dec-2026
289,000
할인액 28,000 (10%)
Each
Thermo Scientific Pierce NBT and BCIP Substrates are powders and ready-made solutions of nitro-blue tetrazolium and 5-bromo-4-chloro-3'-indolyphosphate for chromogenic blot and IHC staining with alkaline phosphatase probes.

The combination of NBT (nitro-blue tetrazolium chloride) and BCIP (5-bromo-4-chloro-3'-indolyphosphate p-toluidine salt) yields an intense, insoluble black-purple precipitate when reacted with alkaline phosphatase, a popular enzyme conjugate for antibody probes. Pierce NBT and BCIP powders are available individually and in two convenient, ready-to-use solutions (with and without a levamisole suppressor of endogenous phosphatase activity).

Features:

• AP substrate—for detection of alkaline phosphatase (alk-phos) activity on solid media, including nitrocellulose and PVDF membranes and fixed tissue samples
• Chromogenic—no special equipment needed for visualization; produces purple-black precipitate that is easy to photograph
• Package options—choose individual NBT and BCIP powders, or preformulated, ready-to-use solutions with or without levamisole suppressor

Highlights of Pierce NBT/BCIP 1-Step Solution (Part No. 34042):
• Best for immunoblotting applications
• Ready-to-use single component
• Sensitive with low background and minimial assay-to-assay variability

Highlights of Pierce NBT/BCIP plus Suppressor 1-Step Solution (Part No. 34070):
• Best for immunohistochemistry applications
• Ready-to-use single component
• Contains 1 mM levamisole to inhibit endogenous phosphatase activity, while yielding an intense dark purple precipitate in the presence of calf intestinal phosphatase

Related Products
1-Step™ NBT/BCIP plus Suppressor Substrate Solution
NBT Substrate Powder (nitro-blue tetrazolium chloride)
BCIP Substrate Powder (5-bromo-4-chloro-3'-indolyphosphate p-toluidine)
For Research Use Only. Not for use in diagnostic procedures.
사양
설명Pierce 1-Step NBT/BCIP Substrate Solution
반응성Alkaline Phosphatase
검출 방법Colorimetric
형태Liquid
Membrane CompatibilityNitrocellulose, PVDF
제품라인1-Step
제품 유형Substrate
수량250 mL
SubstrateAlkaline Phosphatase
Unit SizeEach
구성 및 보관
Upon receipt store product at 4°C. Product is shipped at ambient temperature.

자주 묻는 질문(FAQ)

Will the 1-Step NBT/BCIP Substrate Solution work with PVDF membranes?

Yes, it will work with both PVDF as well as nitrocellulose membranes

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

With the 1-Step NBT/BCIP Substrate Solution, how can I eliminate endogenous phosphatase activity?

To eliminate endogenous phosphatase activity, add a final concentration of 1 mM levamisole to the 1-Step NBT/BCIP Substrate Solution.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

Can I use the 1-Step NBT/BCIP Substrate Solution for ELISA?

No, the 1-Step NBT/BCIP Substrate Solution cannot be used for ELISA. It is a precipitating alkaline phoshpatase substrate developed for immunoblotting, immunohistochemistry and in situ hybridzation. For ELISA, use our 1-Step PNPP Substrate Solution (Cat. No. 37621).

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.