PNPP Substrate Tablets, 105 Tablets - FAQs
View additional product information for PNPP Substrates - FAQs (34047, 34045, 37620, 37621)
2 product FAQs found
Can you provide the shelf life for PNPP Substrate Tablets?
PNPP Substrate Tablets are covered under our general 1-year warranty and are guaranteed to be fully functional for 12 months from the date of shipment, if stored as recommended (-20 degrees C). Please see section 8.1 of our Terms & Conditions of Sale (https://www.thermofisher.com/content/dam/LifeTech/Documents/PDFs/Terms-and-Conditions-of-Sale.pdf).