Nalgene™ PETG Serum Vials; with Crimp Finish: Sterile, Shrink-Wrapped Modules
Nalgene™ PETG Serum Vials; with Crimp Finish: Sterile, Shrink-Wrapped Modules
Nalgene™ PETG Serum Vials; with Crimp Finish: Sterile, Shrink-Wrapped Modules
Nalgene™ PETG Serum Vials; with Crimp Finish: Sterile, Shrink-Wrapped Modules
Nalgene™ PETG Serum Vials; with Crimp Finish: Sterile, Shrink-Wrapped Modules
Thermo Scientific™

Nalgene™ PETG Serum Vials; with Crimp Finish: Sterile, Shrink-Wrapped Modules

Protect against leakage, breakage and contamination by using PETG serum vials with crimp finish. Lighter in weight than glass, these vials are made for use with stoppers and caps.
Catalog NumberVolume (Metric)Closure Size
342030-00033 mL13 mm
342030-00055 mL20 mm
342030-001010 mL20 mm
342030-002020 mL20 mm
Catalog number 342030-0003
Price (JPY)
383,440
Online offer
Ends: 25-Dec-2026
479,300
Save 95,860 (20%)
Case of 3451
Volume (Metric):
3 mL
Closure Size:
13 mm

Protect against leakage, breakage, and contamination. Thermo Scientific™ Nalgene PETG Crimp Finish Serum Vials are lighter in weight than glass, and are made for use with stoppers and caps.

  • PETG
  • Non-cytotoxic
  • Non-pyrogenic
  • Packaged in trayless shrink-wrap modules
  • USP VI-compliant
  • Sterile to 10-6 SAL

Ordering Information:

Serum Vials are packed in trayless shrink-wrapped modules. Stoppers and Caps not included.

A sample product certificate providing Cytotoxicity, USP Class VI, FDA Compliance—21 CFR and other regulatory compliance information is available or can be requested from our Certificates of Compliance Database

Specifications
Certifications/ComplianceISO 13485:2016
Closure Size13 mm
DescriptionSerum Vials
Diameter (Metric) Inner7 mm
For Use With (Equipment)Stopper, Cap
Height (Metric)37 mm
No. per Case3451
No. per Pack493
Product TypeSerum Vial
Volume (English)1/10 oz.
Volume (Metric) Total3 mL
Closure TypeStoppers and Caps (Not Included)
ColorClear
Diameter (Metric) Outer17 mm
MaterialPETG
Product LineNalgene
Quantity3451 Cases, 493 Pk.
SterilitySterile
Volume (Metric)3 mL
Unit SizeCase of 3451