(R)-(+)-3-Pyrolidinol, 98 %, Thermo Scientific Chemicals
(R)-(+)-3-Pyrolidinol, 98 %, Thermo Scientific Chemicals
(R)-(+)-3-Pyrolidinol, 98 %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

(R)-(+)-3-Pyrolidinol, 98 %, Thermo Scientific Chemicals

CAS: 2799-21-5 | C4H9NO | 87.12 g/mol
Have Questions?
Modifier l'affichagebuttonViewtableView
Quantité:
1 g
5 g
Conditionnement:
Flacon en verre
Référence 344570010
Prix (EUR)
52,60
Each
Quantité:
1 g
Conditionnement:
Flacon en verre
Grand volume ou format personnalisé
Prix (EUR)
52,60
Each
Identifiants chimiques
CAS2799-21-5
IUPAC Name(3R)-3-hydroxypyrrolidin-1-ium
Molecular FormulaC4H10NO
InChI KeyJHHZLHWJQPUNKB-SCSAIBSYSA-O
SMILESO[C@@H]1CC[NH2+]C1
Voir plus
Appearance (Color)Clear yellow to orange-brown
Appearance (Form)Viscous liquid
AppearanceMay darken on storage
Infrared spectrumConforms
GC>=97.5 %
Voir plus
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only