1,2,4-Benzenetricarboxylic acid, 98%
1,2,4-Benzenetricarboxylic acid, 98%
1,2,4-Benzenetricarboxylic acid, 98%
Thermo Scientific Chemicals

1,2,4-Benzenetricarboxylic acid, 98%

CAS: 528-44-9 | C9H6O6 | 210.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
50 g
250 g
Packaging:
View all
Glass Bottle
Catalog number 349030050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS528-44-9
IUPAC Namebenzene-1,2,4-tricarboxylic acid
Molecular FormulaC9H6O6
InChI KeyARCGXLSVLAOJQL-UHFFFAOYSA-N
SMILESOC(=O)C1=CC=C(C(O)=O)C(=C1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
HPLC>=97.5 %
Appearance (Form)Crystalline powder
Appearance (Color)White
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only