D-alpha-Tocopherylquinone, 90-95%
D-alpha-Tocopherylquinone, 90-95%
D-alpha-Tocopherylquinone, 90-95%
Thermo Scientific Chemicals

D-alpha-Tocopherylquinone, 90-95%

CAS: 7559-04-8 | C29H50O3 | 446.71 g/mol
Catalog NumberQuantityPackaging
350305000500 mgGlass bottle
Catalog number 350305000
Price (JPY)
-
Quantity:
500 mg
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS7559-04-8
IUPAC Name2-(3-hydroxy-3,7,11,15-tetramethylhexadecyl)-3,5,6-trimethylcyclohexa-2,5-diene-1,4-dione
Molecular FormulaC29H50O3
InChI KeyLTVDFSLWFKLJDQ-UHFFFAOYNA-N
SMILESCC(C)CCCC(C)CCCC(C)CCCC(C)(O)CCC1=C(C)C(=O)C(C)=C(C)C1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear amber to red
Infrared spectrumConforms
Appearance (Form)Viscous liquid
GC>=90 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only