Oxalic acid, potassium salt dihydrate, 99%, extra pure
Oxalic acid, potassium salt dihydrate, 99%, extra pure
Oxalic acid, potassium salt dihydrate, 99%, extra pure
Thermo Scientific Chemicals

Oxalic acid, potassium salt dihydrate, 99%, extra pure

CAS: 6100-20-5 | C4H3KO8 | 218.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
1 kg
Packaging:
Glass bottle
Catalog number 352115000
Price (USD)
-
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS6100-20-5
IUPAC Namepotassium oxalic acid hydrogen oxalate
Molecular FormulaC4H3KO8
InChI KeyGANDVAJEIJXBQJ-UHFFFAOYSA-M
SMILES[K+].OC(=O)C(O)=O.OC(=O)C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to white
Appearance (Form)Crystalline powder or crystals
Infrared spectrumConforms
Titration with NaOH>=98.5 %
Loss on drying12 % to 16 % (1 g, 105°C)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only