1-Ethyl-3-methylimidazolium bromide, 98%
1-Ethyl-3-methylimidazolium bromide, 98%
1-Ethyl-3-methylimidazolium bromide, 98%
Thermo Scientific Chemicals

1-Ethyl-3-methylimidazolium bromide, 98%

CAS: 65039-08-9 | C6H11BrN2 | 191.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 356671000
Price (EUR)
178,00
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
178,00
Each
Chemical Identifiers
CAS65039-08-9
IUPAC Name1-ethyl-3-methyl-1H-imidazol-3-ium bromide
Molecular FormulaC6H11BrN2
InChI KeyGWQYPLXGJIXMMV-UHFFFAOYSA-M
SMILES[Br-].CCN1C=C[N+](C)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow
Titration Argentometric>=97.5 %
Appearance (Form)Crystalline powder or crystals and/or chunks
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Ionic liquid
RUO – Research Use Only