2-Acetylphenylboronic acid, 96%
2-Acetylphenylboronic acid, 96%
2-Acetylphenylboronic acid, 96%
Thermo Scientific Chemicals

2-Acetylphenylboronic acid, 96%

CAS: 308103-40-4 | C8H9BO3 | 163.97 g/mol
Quantity:
1 g
5 g
25 g
Catalog number 358820250
Price (JPY)
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS308103-40-4
IUPAC Name(2-acetylphenyl)boronic acid
Molecular FormulaC8H9BO3
InChI KeyZKAOVABYLXQUTI-UHFFFAOYSA-N
SMILESCC(=O)C1=CC=CC=C1B(O)O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
HPLC>=95 %
Appearance (Color)White to yellow
Appearance (Form)Powder and/or chunks
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only