Potassium tert-butoxide, pure, 20 wt.% solution in THF
Potassium tert-butoxide, pure, 20 wt.% solution in THF
Potassium tert-butoxide, pure, 20 wt.% solution in THF
Thermo Scientific Chemicals

Potassium tert-butoxide, pure, 20 wt.% solution in THF

CAS: 865-47-4 | C4H9KO | 112.21 g/mol
Have Questions?
產品號碼QuantityPackaging
364992500250 mLGlass bottle
產品號碼 364992500
價格 (TWD)
-
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
化學識別
CAS865-47-4
IUPAC Namepotassium 2-methylpropan-2-olate
Molecular FormulaC4H9KO
InChI KeyLPNYRYFBWFDTMA-UHFFFAOYSA-N
SMILES[K+].CC(C)(C)[O-]
檢視更多
規格Specification Sheet規格表
Appearance (Color)Colorless to amber
Appearance (Form)Solution
Strength19 to 21 % (Acidimetric)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only