Zinc chloride, 1.0M solution in diethyl ether, AcroSeal™
Zinc chloride, 1.0M solution in diethyl ether, AcroSeal™
Zinc chloride, 1.0M solution in diethyl ether, AcroSeal™
Zinc chloride, 1.0M solution in diethyl ether, AcroSeal™
Thermo Scientific Chemicals

Zinc chloride, 1.0M solution in diethyl ether, AcroSeal™

CAS: 7646-85-7 | Cl2Zn | 136.28 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
800 mL
Catalog number 370051000
Price (USD)
63.65
Online exclusive
70.30
Save 6.65
Each
Quantity:
100 mL
Request bulk or custom format
Price (USD)
63.65
Online exclusive
70.30
Save 6.65
Each
Chemical Identifiers
CAS7646-85-7
IUPAC Namezinc(2+) dichloride
Molecular FormulaCl2Zn
InChI KeyJIAARYAFYJHUJI-UHFFFAOYSA-L
SMILES[Cl-].[Cl-].[Zn++]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to light yellow to light blue-grey
Appearance (Form)Solution
AppearanceMay develop some turbidity or precipitate
Titration Argentometric0.9 to 1.1 M
Water=<0.2 % (Coulometric)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only