(tert-Butoxycarbonylmethylene)triphenylphosphorane, 97%
(tert-Butoxycarbonylmethylene)triphenylphosphorane, 97%
(tert-Butoxycarbonylmethylene)triphenylphosphorane, 97%
Thermo Scientific Chemicals

(tert-Butoxycarbonylmethylene)triphenylphosphorane, 97%

CAS: 35000-38-5 | C24H25O2P | 376.44 g/mol
Have Questions?
Catalog NumberQuantityPackaging
37062025025 gGlass bottle
Catalog number 370620250
Price (USD)
235.65
Special offer
Online exclusive
Ends: 28-Dec-2026
277.00
Save 41.35
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
235.65
Special offer
Online exclusive
Ends: 28-Dec-2026
277.00
Save 41.35
Each
Chemical Identifiers
CAS35000-38-5
IUPAC Nametert-butyl 2-(triphenyl-λ⁵-phosphanylidene)acetate
Molecular FormulaC24H25O2P
InChI KeyZWZUFQPXYVYAFO-UHFFFAOYSA-N
SMILESCC(C)(C)OC(=O)C=P(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Carbon (C)73.5 to 79.5 %
Appearance (Color)White to yellow
Infrared spectrumConforms
Melting point146°C to 155°C
Appearance (Form)Powder or crystals
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For C-C Bond Formation (Olefination)
RUO – Research Use Only