Formic acid hydrazide, 90%
Formic acid hydrazide, 90%
Formic acid hydrazide, 90%
Thermo Scientific Chemicals

Formic acid hydrazide, 90%

CAS: 624-84-0 | CH4N2O | 60.06 g/mol
Have Questions?
Catalog NumberQuantityPackaging
37606025025 gGlass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 376060250
Price (USD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS624-84-0
IUPAC Nameformohydrazide
Molecular FormulaCH4N2O
InChI KeyXZBIXDPGRMLSTC-UHFFFAOYSA-N
SMILESNNC=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)White to pink
Appearance (Form)Crystalline powder or flakes
Melting point51°C to 59°C
Titration with KIO388 to 112 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only