4-Fluorophenylacetyl chloride, 98%
4-Fluorophenylacetyl chloride, 98%
4-Fluorophenylacetyl chloride, 98%
Thermo Scientific Chemicals

4-Fluorophenylacetyl chloride, 98%

CAS: 459-04-1 | C8H6ClFO | 172.58 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Catalog number 376120050
Price (EUR)
127,00
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
127,00
Each
Chemical Identifiers
CAS459-04-1
IUPAC Name2-(4-fluorophenyl)acetyl chloride
Molecular FormulaC8H6ClFO
InChI KeySIOJFYRPBYGHOO-UHFFFAOYSA-N
SMILESFC1=CC=C(CC(Cl)=O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.5110 to 1.5130 (20°C, 589 nm)
Infrared spectrumConforms
Appearance (Form)Liquid
GC>=97.5 %
Appearance (Color)Clear yellow
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only