Product Image
Thermo Scientific Chemicals

Benzaldehyde, 99.5+%, pure, redistilled, AcroSeal™

CAS: 100-52-7 | C7H6O | 106.12 g/mol
Quantity:
100 mL
1 L
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 378361000
Price (JPY)
-
Quantity:
100 mL
Request bulk or custom format
Chemical Identifiers
CAS100-52-7
IUPAC Namebenzaldehyde
Molecular FormulaC7H6O
InChI KeyHUMNYLRZRPPJDN-UHFFFAOYSA-N
SMILESO=CC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
GC>=99.50 %
Refractive index1.5440 to 1.5460 (20°C, 589 nm)
Infrared spectrumConforms
Appearance (Color)Clear colorless
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only