2-Methylindole-3-carboxaldehyde, 97+%, Thermo Scientific Chemicals
2-Methylindole-3-carboxaldehyde, 97+%, Thermo Scientific Chemicals
2-Methylindole-3-carboxaldehyde, 97+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Methylindole-3-carboxaldehyde, 97+%, Thermo Scientific Chemicals

数量:
25 g
100 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 379720250
価格(JPY)
-
数量:
25 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS5416-80-8
IUPAC Name2-methyl-1H-indole-3-carbaldehyde
Molecular FormulaC10H9NO
InChI KeyCYZIVXOEJNAIBS-UHFFFAOYSA-N
SMILESCC1=C(C=O)C2=CC=CC=C2N1
さらに表示
HPLC>=97.0 %
Appearance (Color)Brown
Appearance (Form)Powder
Infrared spectrumConforms
Melting point199°C to 202°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only