Zinc chloride, 99.99%, (trace metal basis), anhydrous
Zinc chloride, 99.99%, (trace metal basis), anhydrous
Zinc chloride, 99.99%, (trace metal basis), anhydrous
Thermo Scientific Chemicals

Zinc chloride, 99.99%, (trace metal basis), anhydrous

CAS: 7646-85-7 | Cl2Zn | 136.29 g/mol
製品番号(カタログ番号) 380130250
価格(JPY)
-
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS7646-85-7
IUPAC Namezinc(2+) dichloride
Molecular FormulaCl2Zn
InChI KeyJIAARYAFYJHUJI-UHFFFAOYSA-L
SMILES[Cl-].[Cl-].[Zn++]
さらに表示
Appearance (Form)Crystalline powder or crystals
Appearance (Color)White to light grey
Total trace metal impurities=<0.01 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysis
RUO – Research Use Only