Bis(norbornadiene)rhodium(I) tetrafluoroborate, 94%
Bis(norbornadiene)rhodium(I) tetrafluoroborate, 94%
Bis(norbornadiene)rhodium(I) tetrafluoroborate, 94%
Thermo Scientific Chemicals

Bis(norbornadiene)rhodium(I) tetrafluoroborate, 94%

CAS: 36620-11-8 | C14H16BF4Rh | 373.99 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mg
500 mg
Packaging:
Glass bottle
Catalog number 382551000
Price (USD)
-
Quantity:
100 mg
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS36620-11-8
IUPAC Nameλ¹-rhodium(1+) bis(bicyclo[2.2.1]hepta-2,5-diene) tetrafluoroboranuide
Molecular FormulaC14H16BF4Rh
InChI KeyHAYDJWBQWOEERB-UHFFFAOYSA-N
SMILES[Rh+].F[B-](F)(F)F.C1C2C=CC1C=C2.C1C2C=CC1C=C2
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Red
Appearance (Form)Powder
Infrared spectrumConforms
Elemental Analysis>=92 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only