Di-t-butylmethylphosphonium tetrafluoroborate
Di-t-butylmethylphosphonium tetrafluoroborate
Di-t-butylmethylphosphonium tetrafluoroborate
Thermo Scientific Chemicals

Di-t-butylmethylphosphonium tetrafluoroborate

CAS: 479094-62-7 | C9H22BF4P | 248.05 g/mol
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 386960010
Price (JPY)
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS479094-62-7
IUPAC Namedi-tert-butyl(methyl)phosphanium; tetrafluoroboranuide
Molecular FormulaC9H22BF4P
InChI KeyBRDLRXCAHKUWJS-UHFFFAOYSA-O
SMILESF[B-](F)(F)F.C[PH+](C(C)(C)C)C(C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
NMR (31P-NMR)>=99
Appearance (Form)Crystalline powder
NMR (1H-NMR)Conforms
Appearance (Color)White to light yellow
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only