Boron trifluoride dihydrate, 65% BF3
Boron trifluoride dihydrate, 65% BF3
Boron trifluoride dihydrate, 65% BF3
Thermo Scientific Chemicals

Boron trifluoride dihydrate, 65% BF3

CAS: 13319-75-0 | BF3H4O2 | 103.84 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
Packaging:
Glass bottle
Catalog number 388711000
Price (USD)
-
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS13319-75-0
IUPAC Nametrifluoroborane dihydrate
Molecular FormulaBF3H4O2
InChI KeyMJCYPBSRKLJZTB-UHFFFAOYSA-N
SMILESO.O.FB(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Appearance (Color)Clear colorless to light yellow
Assay65.0 to 66.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lewis acid
RUO – Research Use Only