1-Octanesulfonic acid, sodium salt, 99+%, Ion pair chromatography, anhydrous
1-Octanesulfonic acid, sodium salt, 99+%, Ion pair chromatography, anhydrous
1-Octanesulfonic acid, sodium salt, 99+%, Ion pair chromatography, anhydrous
Thermo Scientific Chemicals

1-Octanesulfonic acid, sodium salt, 99+%, Ion pair chromatography, anhydrous

CAS: 5324-84-5 | C8H17NaO3S | 216.27 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 396690250
Price (USD)
333.65
Special offer
Online exclusive
Ends: 28-Dec-2026
392.00
Save 58.35
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
333.65
Special offer
Online exclusive
Ends: 28-Dec-2026
392.00
Save 58.35
Each
Chemical Identifiers
CAS5324-84-5
IUPAC Namesodium octane-1-sulfonate
Molecular FormulaC8H17NaO3S
InChI KeyHRQDCDQDOPSGBR-UHFFFAOYSA-M
SMILES[Na+].CCCCCCCCS([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Loss on drying=<2 % (150°C)
UVat 210 nm A: =<0.1
Infrared spectrumConforms
UV(0.4 M in water)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ion pair chromatography
RUO – Research Use Only