3-Chloro-2,4(3H,5H)-furandione, 97%
3-Chloro-2,4(3H,5H)-furandione, 97%
3-Chloro-2,4(3H,5H)-furandione, 97%
Thermo Scientific Chemicals

3-Chloro-2,4(3H,5H)-furandione, 97%

CAS: 4971-55-5 | C4H3ClO3 | 134.52 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number 399650100
Price (EUR)
38,00
Each
Quantity:
10 g
Request bulk or custom format
Price (EUR)
38,00
Each
Chemical Identifiers
CAS4971-55-5
IUPAC Name3-chlorooxolane-2,4-dione
Molecular FormulaC4H3ClO3
InChI KeyDJEJYZZJYYNADA-UHFFFAOYNA-N
SMILESClC1C(=O)COC1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Beige
HPLC>=96.0 %
Appearance (Form)Crystalline powder
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only