Alcohol 4-aminofenetilo, 97 %, Thermo Scientific Chemicals
Alcohol 4-aminofenetilo, 97 %, Thermo Scientific Chemicals
Alcohol 4-aminofenetilo, 97 %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Alcohol 4-aminofenetilo, 97 %, Thermo Scientific Chemicals

CAS: 104-10-9 | C8H11NO | 137.18 g/mol
Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
25 g
100 g
Envase:
Frasco de vidrio
Número de catálogo 400931000
Precio (USD)
-
Cantidad:
100 g
Envase:
Frasco de vidrio
Pedido a granel o personalizado
Identificadores químicos
CAS104-10-9
IUPAC Name2-(4-aminophenyl)ethan-1-ol
Molecular FormulaC8H11NO
InChI KeyQXHDYMUPPXAMPQ-UHFFFAOYSA-N
SMILESNC1=CC=C(CCO)C=C1
Ver más
EspecificacionesSpecification SheetHoja de especificaciones
GC>=96.0 %
Appearance (Color)Off-white to brown
Appearance (Form)Crystalline powder or crystals
Infrared spectrumConforms
Melting point106°C to 110°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only