p-Anisidine hydrochloride, 99%
p-Anisidine hydrochloride, 99%
p-Anisidine hydrochloride, 99%
Thermo Scientific Chemicals

p-Anisidine hydrochloride, 99%

CAS: 20265-97-8 | C7H9NO·HCl | 159.61 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number 401300250
Price (USD)
111.65
Special offer
Online exclusive
Ends: 28-Dec-2026
131.00
Save 19.35
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
111.65
Special offer
Online exclusive
Ends: 28-Dec-2026
131.00
Save 19.35
Each
Chemical Identifiers
CAS20265-97-8
IUPAC Namehydrogen 4-methoxyaniline chloride
Molecular FormulaC7H10ClNO
InChI KeyVQYJLACQFYZHCO-UHFFFAOYSA-N
SMILES[H+].[Cl-].COC1=CC=C(N)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Off-white to pink to grey-brown
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Melting point214°C to 219°C
Titration with HClO4>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only