CD4 Monoclonal Antibody (4SM95), eFluor™ 570, eBioscience™ - FAQs
CD4 Monoclonal Antibody (4SM95), eFluor™ 570, eBioscience™ - FAQs
View additional product information for CD4 Monoclonal Antibody (4SM95), eFluor™ 570, eBioscience™ - FAQs (41-9766-80)