Ethyl oleate, 98%, mixture of homologeous fatty acid esters
Ethyl oleate, 98%, mixture of homologeous fatty acid esters
Ethyl oleate, 98%, mixture of homologeous fatty acid esters
Thermo Scientific Chemicals

Ethyl oleate, 98%, mixture of homologeous fatty acid esters

CAS: 111-62-6 | C20H38O2 | 310.51 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
250 g
1 kg
2.5 kg
Packaging:
檢視全部
Glass bottle
產品號碼 410292500
價格 (TWD)
-
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
化學識別
CAS111-62-6
IUPAC Nameethyl (9E)-octadec-9-enoate
Molecular FormulaC20H38O2
InChI KeyLVGKNOAMLMIIKO-VAWYXSNFSA-N
SMILESCCCCCCCC\C=C\CCCCCCCC(=O)OCC
檢視更多
規格Specification Sheet規格表
Infrared spectrumConforms
Saponification value177 to 188 mg KOH/g
Water=<1 % (K.F.)
Appearance (Form)Clear viscous liquid
Acid value=<0.5 mg KOH/g
檢視更多
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only