2-Methyl-1-propanol, 99+%, ACS reagent
2-Methyl-1-propanol, 99+%, ACS reagent
2-Methyl-1-propanol, 99+%, ACS reagent
Thermo Scientific Chemicals

2-Methyl-1-propanol, 99+%, ACS reagent

CAS: 78-83-1 | C4H10O | 74.12 g/mol
Quantity:
500 mL
2.5 L
Packaging:
View all
Glass Bottle
Catalog number 412655000
Price (JPY)
-
Quantity:
500 mL
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS78-83-1
IUPAC Name2-methylpropan-1-ol
Molecular FormulaC4H10O
InChI KeyZXEKIIBDNHEJCQ-UHFFFAOYSA-N
SMILESCC(C)CO
View more
SpecificationsSpecification SheetSpecification Sheet
Color scale=<10 APHA
Infrared spectrumConforms
GC>=99.0 %
Water=<0.1 % (K.F.)
Solubility(in water) Passes test
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only