2-Methyl-2-butene, 90%, balance 2-Methyl-1-butene
2-Methyl-2-butene, 90%, balance 2-Methyl-1-butene
2-Methyl-2-butene, 90%, balance 2-Methyl-1-butene
Thermo Scientific Chemicals

2-Methyl-2-butene, 90%, balance 2-Methyl-1-butene

CAS: 513-35-9 | C5H10 | 70.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
2.5 L
Packaging:
Glass bottle
Catalog number 414090010
Price (USD)
-
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS513-35-9
IUPAC Name2-methylbut-2-ene
Molecular FormulaC5H10
InChI KeyBKOOMYPCSUNDGP-UHFFFAOYSA-N
SMILESCC=C(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
GCBalance mainly 2-methyl-1-butene
GC>=88 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only