Methyl glycolate, 98%
Methyl glycolate, 98%
Methyl glycolate, 98%
Thermo Scientific Chemicals

Methyl glycolate, 98%

CAS: 96-35-5 | C3H6O3 | 90.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 414371000
Price (EUR)
377,00
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
377,00
Each
Chemical Identifiers
CAS96-35-5
IUPAC Namemethyl 2-hydroxyacetate
Molecular FormulaC3H6O3
InChI KeyGSJFXBNYJCXDGI-UHFFFAOYSA-N
SMILESCOC(=O)CO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Appearance (Color)Clear colorless
Refractive index1.4150 to 1.4180 (20°C, 589 nm)
GC>=97.5 %
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only