Methyl valerate, 99%
Methyl valerate, 99%
Methyl valerate, 99%
Thermo Scientific Chemicals

Methyl valerate, 99%

CAS: 624-24-8 | C6H12O2 | 116.16 g/mol
Have Questions?
Catalog NumberQuantity
41505025025 g
Catalog number 415050250
Price (EUR)
33,65
Online exclusive
37,60
Save 3,95 (11%)
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
33,65
Online exclusive
37,60
Save 3,95 (11%)
Each
Chemical Identifiers
CAS624-24-8
IUPAC Namemethyl pentanoate
Molecular FormulaC6H12O2
InChI KeyHNBDRPTVWVGKBR-UHFFFAOYSA-N
SMILESCCCCC(=O)OC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=98.5 %
Refractive index1.3965 to 1.3985 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only