Potassium chlorate, 99+%, ACS reagent
Potassium chlorate, 99+%, ACS reagent
Potassium chlorate, 99+%, ACS reagent
Thermo Scientific Chemicals

Potassium chlorate, 99+%, ACS reagent

CAS: 3811-04-9 | ClKO3 | 122.55 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
500 g
2.5 kg
Packaging:
View all
Plastic bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 418190025
Price (USD)
-
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS3811-04-9
IUPAC Namepotassium chlorate
Molecular FormulaClKO3
InChI KeyVKJKEPKFPUWCAS-UHFFFAOYSA-M
SMILES[K+].[O-][Cl](=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Chloride (Cl)=<10 ppm
Heavy metals (as Pb)=<5 ppm
Appearance (Form)Crystalline powder
Sulfate (SO4)Passes test (limit about 0.002%)
Iron (Fe)=<3 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent
RUO – Research Use Only