Potassium bis(trimethylsilyl)amide, 0.7M (15 wt.%) solution in toluene, AcroSeal™
Potassium bis(trimethylsilyl)amide, 0.7M (15 wt.%) solution in toluene, AcroSeal™
Potassium bis(trimethylsilyl)amide, 0.7M (15 wt.%) solution in toluene, AcroSeal™
Thermo Scientific Chemicals

Potassium bis(trimethylsilyl)amide, 0.7M (15 wt.%) solution in toluene, AcroSeal™

CAS: 40949-94-8 | C6H18KNSi2 | 199.485 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
800 mL
Packaging:
View all
AcroSeal™ Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 418231000
Price (USD)
92.10
Each
Quantity:
100 mL
Packaging:
AcroSeal™ Glass bottle
Request bulk or custom format
Price (USD)
92.10
Each
Chemical Identifiers
CAS40949-94-8
IUPAC Namepotassium bis(trimethylsilyl)azanide
Molecular FormulaC6H18KNSi2
InChI KeyIUBQJLUDMLPAGT-UHFFFAOYSA-N
SMILES[K+].C[Si](C)(C)[N-][Si](C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Titration with HClO40.6 to 0.9 M
Appearance (Form)Solution
Appearance (Color)Clear to slightly hazy colorless to amber
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only