L(+)-Prolinol, 95%
L(+)-Prolinol, 95%
L(+)-Prolinol, 95%
Thermo Scientific Chemicals

L(+)-Prolinol, 95%

CAS: 23356-96-9 | C5H11NO | 101.149 g/mol
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 418320250
Price (JPY)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS23356-96-9
IUPAC Name(pyrrolidin-2-yl)methanol
Molecular FormulaC5H11NO
InChI KeyHVVNJUAVDAZWCB-UHFFFAOYNA-N
SMILESOCC1CCCN1
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.4840 to 1.4870 (20°C, 589 nm)
Specific optical rotation+29° to +33° (20°C, 589 nm) (c=1, toluene)
Appearance (Color)Clear colorless to yellow
Appearance (Form)Liquid
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only