Tetraethylammonium Hydroxide, 10% in Water
Tetraethylammonium Hydroxide, 10% in Water
Tetraethylammonium Hydroxide, 10% in Water
Thermo Scientific Chemicals

Tetraethylammonium Hydroxide, 10% in Water

CAS: 77-98-5 | C8H21NO | 147.26 g/mol
Quantity:
100 g
500 g
Catalog number 420281000
Price (JPY)
-
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS77-98-5
IUPAC Nametetraethylazanium hydroxide
Molecular FormulaC8H21NO
InChI KeyLRGJRHZIDJQFCL-UHFFFAOYSA-M
SMILES[OH-].CC[N+](CC)(CC)CC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Appearance (Form)Solution
Titration with HCl9.5 to 10.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Phase transfer catalyst
RUO – Research Use Only