2,2,2-Tribromoethanol, 99%
2,2,2-Tribromoethanol, 99%
2,2,2-Tribromoethanol, 99%
Thermo Scientific Chemicals

2,2,2-Tribromoethanol, 99%

CAS: 75-80-9 | C2H3Br3O | 282.76 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Packaging:
Glass bottle
Catalog number 421430100
Price (EUR)
43,60
Each
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
43,60
Each
Chemical Identifiers
CAS75-80-9
IUPAC Name2,2,2-tribromoethan-1-ol
Molecular FormulaC2H3Br3O
InChI KeyYFDSDPIBEUFTMI-UHFFFAOYSA-N
SMILESOCC(Br)(Br)Br
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=98.5 %
Appearance (Color)White to yellow to light brown
Infrared spectrumConforms
Appearance (Form)Crystalline powder or crystals
Melting point73°C to 82°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only