1,1,2-Trimethyl-1H-benz[e]indole, 96%
1,1,2-Trimethyl-1H-benz[e]indole, 96%
1,1,2-Trimethyl-1H-benz[e]indole, 96%
Thermo Scientific Chemicals

1,1,2-Trimethyl-1H-benz[e]indole, 96%

CAS: 41532-84-7 | C15H15N | 209.29 g/mol
Have Questions?
Catalog NumberQuantity
42190025025 g
Catalog number 421900250
Price (USD)
88.65
Special offer
Online exclusive
Ends: 28-Dec-2026
104.00
Save 15.35
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
88.65
Special offer
Online exclusive
Ends: 28-Dec-2026
104.00
Save 15.35
Each
Chemical Identifiers
CAS41532-84-7
IUPAC Name1,1,2-trimethyl-1H-benzo[e]indole
Molecular FormulaC15H15N
InChI KeyWJZSZXCWMATYFX-UHFFFAOYSA-N
SMILESCC1=NC2=CC=C3C=CC=CC3=C2C1(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Melting point111°C to 117°C
HPLC>=95 %
Appearance (Color)Yellow to brown
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only