Matrix™ DeepWell™ Storage Blocks, 96 well, 2mL
Actual product may vary
Thermo Scientific™

Matrix™ DeepWell™ Storage Blocks, 96 well, 2mL

Use these DeepWell™ storage blocks as mother plates when creating compound libraries from automated liquid handling instruments.
Catalog number 4221
Price (JPY)
50,880
Case of 40

Use Thermo Scientific™ Matrix™ DeepWell™ Storage Blocks as mother plates when creating compound libraries from automated liquid handling instruments. They are constructed of polypropylene, are chemically resistant and meet all standard microplate specifications. The unique chimney well design of the 1mL block reduces risk of cross-contamination between wells during pipetting, while the 2mL block has cut-away sides for easy handling and placement on robotic systems.

Features:

  • Alphanumeric notations simplify the identification of wells
  • Industry-standard footprint ensures compatibility with sealers, stackers and liquid handlers
Specifications
AutoclavableAutoclavable
Binding TypeLow binding PP
Certifications/ComplianceCertified RNase and DNase Free. ANSI Compliant.
For Use With (Equipment)ALPS Heat Sealers, Multidrop Combi, Centrifuges, Ultralow Freezers, Liquid Handlers
FormatStandard
No. per Case40
No. per Pack40
Packaging40 Cases
Product TypePolypropylene Storage & Sample Prep Plate
Regulatory StatusRUO
Volume (Metric) Well2200 μL
Volume (Metric) Working2000 μL
Array8 x 12
BarcodeCustom Barcoding Available
ColorNatural
MaterialPolypropylene
No. of Wells96
Product LineNunc
SterilityNon-sterile
Surface TreatmentNon-treated
Well ShapeRound
Unit SizeCase of 40