Tyloxapol, Thermo Scientific Chemicals
Tyloxapol, Thermo Scientific Chemicals
Tyloxapol, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tyloxapol, Thermo Scientific Chemicals

Have Questions?
RéférenceQuantité
4223700505 g
Référence 422370050
Prix (EUR)
36,65
Offre exceptionnelle en ligne
40,50
Économisez 3,85 (10%)
Each
Quantité:
5 g
Grand volume ou format personnalisé
Prix (EUR)
36,65
Offre exceptionnelle en ligne
40,50
Économisez 3,85 (10%)
Each
Identifiants chimiques
CAS25301-02-4
Infrared spectrumConforms
Appearance (Form)Viscous liquid
Appearance (Color)Clear to hazy yellow to amber
pH4.0 to 7.0 (5 % in water)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only