Cobalt(II) nitrate hexahydrate, 98+%, ACS reagent
Cobalt(II) nitrate hexahydrate, 98+%, ACS reagent
Cobalt(II) nitrate hexahydrate, 98+%, ACS reagent
Thermo Scientific Chemicals

Cobalt(II) nitrate hexahydrate, 98+%, ACS reagent

CAS: 10026-22-9 | CoH12N2O12 | 291.03 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
500 g
2.5 kg
Packaging:
View all
Plastic bottle
Catalog number 423585000
Price (USD)
290.65
Online Exclusive
323.00
Save 32.35 (10%)
Each
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
290.65
Online Exclusive
323.00
Save 32.35 (10%)
Each
Chemical Identifiers
CAS10026-22-9
IUPAC Nameλ²-cobalt(2+) hexahydrate dinitrate
Molecular FormulaCoH12N2O12
InChI KeyQGUAJWGNOXCYJF-UHFFFAOYSA-N
SMILESO.O.O.O.O.O.[Co++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Sulfate (SO4)=<0.005 %
Calcium (Ca)=<0.005 %
Lead (Pb)=<0.002 %
Copper (Cu)=<0.002 %
Nickel (Ni)=<0.15 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only