Sodium chloride, 99+%, ACS reagent
Sodium chloride, 99+%, ACS reagent
Sodium chloride, 99+%, ACS reagent
Thermo Scientific Chemicals

Sodium chloride, 99+%, ACS reagent

CAS: 7647-14-5 | ClNa | 58.44 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
500 g
1 kg
5 kg
25 kg
Packaging:
View all
Amber Glass Bottle
Catalog number 424290010
Price (USD)
96.65
Special offer
Online exclusive
Ends: 28-Dec-2026
114.00
Save 17.35
Each
Quantity:
1 kg
Packaging:
Amber Glass Bottle
Request bulk or custom format
Price (USD)
96.65
Special offer
Online exclusive
Ends: 28-Dec-2026
114.00
Save 17.35
Each
Chemical Identifiers
CAS7647-14-5
IUPAC Namesodium chloride
Molecular FormulaClNa
InChI KeyFAPWRFPIFSIZLT-UHFFFAOYSA-M
SMILES[Na+].[Cl-]
View more
SpecificationsSpecification SheetSpecification Sheet
Magnesium (Mg)=<0.001 %
Potassium (K)=<0.005 %
Heavy metals (ICP-OES)=<5 ppm
pH5.0 to 9.0 (5 % solution at 25.0°C)
Insoluble matter=<0.005 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only