Potassium tert-butoxide, 1M solution in tert-butanol, AcroSeal™
Potassium tert-butoxide, 1M solution in tert-butanol, AcroSeal™
Potassium tert-butoxide, 1M solution in tert-butanol, AcroSeal™
Potassium tert-butoxide, 1M solution in tert-butanol, AcroSeal™
Thermo Scientific Chemicals

Potassium tert-butoxide, 1M solution in tert-butanol, AcroSeal™

CAS: 865-47-4 | C4H9KO | 112.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
800 mL
Packaging:
View all
AcroSeal™ Glass Bottle
Catalog number 426121000
Price (USD)
96.00
Each
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (USD)
96.00
Each
Chemical Identifiers
CAS865-47-4
IUPAC Namepotassium 2-methylpropan-2-olate
Molecular FormulaC4H9KO
InChI KeyLPNYRYFBWFDTMA-UHFFFAOYSA-N
SMILES[K+].CC(C)(C)[O-]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to amber
Appearance (Form)Solution
Strength0.95 to 1.10 M
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only