Nalgene™ Polypropylene Copolymer (PPCO) Separatory Funnels with Closure
Nalgene™ Polypropylene Copolymer (PPCO) Separatory Funnels with Closure
Nalgene™ Polypropylene Copolymer (PPCO) Separatory Funnels with Closure
Nalgene™ Polypropylene Copolymer (PPCO) Separatory Funnels with Closure
Thermo Scientific™

Nalgene™ Polypropylene Copolymer (PPCO) Separatory Funnels with Closure

Perform a variety of lab duties with chemically inert, translucent and autoclavable polypropylene separatory funnels.
Catalog NumberCapacity (Metric)
4300-0500PK
also known as 4300-0500
500 mL
4300-0125PK
also known as 4300-0125
125 mL
4300-0250PK
also known as 4300-0250
250 mL
4300-1000PK
also known as 4300-1000
1000 mL
Catalog number 4300-0500PK
also known as 4300-0500
Price (JPY)
16,940
Each
Capacity (Metric):
500 mL

Perform a variety of liquid phase separations with translucent and autoclavable Thermo Scientific™ Nalgene™ Polypropylene Copolymer (PPCO) Separatory Funnels. Good chemical compatibility with a wide range of laboratory solutions involved in liquid-liquid extractions. Polypropylene closure and TFE stopcock require no lubrication. These separatory funnels are impact resistant and won't chip or shatter, making them a valuable asset to lab safety programs.

Features:

  • Separation lines between immiscible liquids are easily observed
  • Phase interface is observable down to the stopcock
  • TFE stopcocks need no lubrication, allowing for easier handling and operation
  • Funnel is chemically compatible with a wide range of lab reagents
  • Autoclavable/Leakproof† when used with Nalgene closures

Includes:

  • Polypropylene funnel, TFE stopcock and housing, polypropylene screw closure
  • Replacement closures also available

Ordering Information:

  • For replacement stopcock, order Cat. No. 714301-1025

Notes:

  • Funnels are susceptible to softening and swelling from extended exposure to aggressive solvents though the effect is reversible.
  • Stem lengths, 65mm.
  • Autoclavable except for stopcock assembly, which is easily removed and chemically sterilized.

Recycling Information:
Nalgene reusable labware products are designed to last many years under typical laboratory conditions. Yet, as with all durable products, they need to be retired and replaced eventually. Thermo Fisher Scientific encourages recycling Nalgene polypropylene (PP) and polypropylene copolymer (PPCO) labware at end of product life in communities where polypropylene can be recycled. Please also recycle the corrugate cartons and any plastic bags (LDPE) included in your product packaging.

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

Specifications
Description43mm closure
Quantity1/Pk.; 4/Cs.
Capacity (Metric)500 mL
Height (Metric)347 mm
MaterialPolypropylene Copolymer (PPCO)
Product TypeSeparatory Funnel
Unit SizeEach