FMOC-Cys(tBu)-OH, 98%
FMOC-Cys(tBu)-OH, 98%
FMOC-Cys(tBu)-OH, 98%
Thermo Scientific Chemicals

FMOC-Cys(tBu)-OH, 98%

CAS: 67436-13-9 | C22H25NO4S | 399.51 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 431270050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS67436-13-9
IUPAC Name3-(tert-butylsulfanyl)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)propanoic acid
Molecular FormulaC22H25NO4S
InChI KeyIXAYZHCPEYTWHW-UHFFFAOYNA-N
SMILESCC(C)(C)SCC(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Specific optical rotation-21.0° to -23.0° (20°C, 589 nm) (c=1, DMF)
Melting point136°C to 140°C
Appearance (Color)White
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only