4-Amino-2-hydroxypyridine, 97%
4-Amino-2-hydroxypyridine, 97%
4-Amino-2-hydroxypyridine, 97%
Thermo Scientific Chemicals

4-Amino-2-hydroxypyridine, 97%

CAS: 38767-72-5 | C5H6N2O | 110.12 g/mol
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 432080010
Price (JPY)
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS38767-72-5
IUPAC Name4-amino-1,2-dihydropyridin-2-one
Molecular FormulaC5H6N2O
InChI KeySBQVQYPJWLJRQT-UHFFFAOYSA-N
SMILESNC1=CC(=O)NC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Powder
HPLC>=96.0 %
Appearance (Color)White to yellow to green to brown
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only