4-tert-Butylphenylmagnesium bromide, 2M solution in diethyl ether, AcroSeal™
4-tert-Butylphenylmagnesium bromide, 2M solution in diethyl ether, AcroSeal™
4-tert-Butylphenylmagnesium bromide, 2M solution in diethyl ether, AcroSeal™
4-tert-Butylphenylmagnesium bromide, 2M solution in diethyl ether, AcroSeal™
Thermo Scientific Chemicals

4-tert-Butylphenylmagnesium bromide, 2M solution in diethyl ether, AcroSeal™

CAS: 63488-10-8 | C10H13BrMg | 237.42 g/mol
Have Questions?
Catalog NumberQuantityPackaging
432861000100 mLAcroSeal™ Glass Bottle
Catalog number 432861000
Price (USD)
265.65
Special offer
Online exclusive
Ends: 28-Dec-2026
312.00
Save 46.35
Each
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (USD)
265.65
Special offer
Online exclusive
Ends: 28-Dec-2026
312.00
Save 46.35
Each
Chemical Identifiers
CAS63488-10-8
IUPAC Namebromo(4-tert-butylphenyl)magnesium
Molecular FormulaC10H13BrMg
InChI KeyYECFWFOEQLAVNH-UHFFFAOYSA-M
SMILESCC(C)(C)C1=CC=C([Mg]Br)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to orange to brown
Titration Grignard1.50 to 2.10 M
Appearance (Form)Solution
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only