4-Aminopiperidine, 95%
4-Aminopiperidine, 95%
4-Aminopiperidine, 95%
Thermo Scientific Chemicals

4-Aminopiperidine, 95%

CAS: 13035-19-3 | C5H12N2 | 100.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 432920250
Price (USD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS13035-19-3
IUPAC Namepiperidin-4-amine
Molecular FormulaC5H12N2
InChI KeyBCIIMDOZSUCSEN-UHFFFAOYSA-N
SMILESNC1CCNCC1
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=94.0 %
Appearance (Form)Liquid or low melting solid
Appearance (Color)Colorless to yellow to brown
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only