Methyl 6-chloropyridazine-4-carboxylate, 97%
Methyl 6-chloropyridazine-4-carboxylate, 97%
Methyl 6-chloropyridazine-4-carboxylate, 97%
Thermo Scientific Chemicals

Methyl 6-chloropyridazine-4-carboxylate, 97%

CAS: 1093860-48-0 | C6H5ClN2O2 | 172.57 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 432980010
Price (USD)
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS1093860-48-0
IUPAC Namemethyl 6-chloropyridazine-4-carboxylate
Molecular FormulaC6H5ClN2O2
InChI KeyQYQHGLKSLGUINZ-UHFFFAOYSA-N
SMILESCOC(=O)C1=CC(Cl)=NN=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point81°C to 89°C
Appearance (Form)Crystalline powder or solid
Appearance (Color)Cream to brown
Infrared spectrumConforms
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only