2-Aminoethanethiol, 95%
2-Aminoethanethiol, 95%
2-Aminoethanethiol, 95%
Thermo Scientific Chemicals

2-Aminoethanethiol, 95%

CAS: 60-23-1 | C2H7NS | 77.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 434710050
Price (USD)
83.65
Special offer
Online exclusive
Ends: 28-Dec-2026
98.00
Save 14.35
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
83.65
Special offer
Online exclusive
Ends: 28-Dec-2026
98.00
Save 14.35
Each
Chemical Identifiers
CAS60-23-1
IUPAC Name2-aminoethane-1-thiol
Molecular FormulaC2H7NS
InChI KeyUFULAYFCSOUIOV-UHFFFAOYSA-N
SMILESNCCS
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow
Infrared spectrumConforms
Titration Iodimetric>=94 %
Appearance (Form)Powder or crystals
Melting point94°C to 99°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only