Gold, 99.99%, (trace metal basis), shot
Gold, 99.99%, (trace metal basis), shot
Gold, 99.99%, (trace metal basis), shot
Thermo Scientific Chemicals

Gold, 99.99%, (trace metal basis), shot

CAS: 7440-57-5 | Au | 196.97 g/mol
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 437140050
Price (JPY)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS7440-57-5
IUPAC Namegold
Molecular FormulaAu
InChI KeyPCHJSUWPFVWCPO-UHFFFAOYSA-N
SMILES[Au]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Beads or pellets
Appearance (Color)Golden
Total trace metal impurities=<100 ppm
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for ultra trace analysis
RUO – Research Use Only